missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chloride Standard, 10 ppm Cl-, Ricca Chemical
Supplier: Ricca Chemical Company R19421004A
Specifications
CHLORIDE STANDARD | |
ClH4N | |
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
N.Cl | |
53.49 | |
CHEBI:31206 | |
4 L |
12125-02-9 | |
MFCD00011420 | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
amine hydrochloride | |
25517 | |
Natural Poly Bottle |