Please login to your online account to display your discounted pricing

Alfa Aesar™ Chloride, Ion chromatography standard solution, Specpure™, Cl- 1000μg/mL

$157.00 - $157.00

Chemical Identifiers

Chemical Name or Material Chloride Ion chromatography standard solution
Name Note Cl- 1000μg/mL
Purity Grade Specpure™
MDL Number MFCD00143949
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA35551AE

 
alfa aesar™
35551AE
100mL Each for $157.00

AA35551AP

 
alfa aesar™
35551AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Chloride Ion chromatography standard solution
Quantity 100mL
TSCA TSCA
Name Note Cl- 1000μg/mL
Purity Grade Specpure™
Physical Form Liquid
MDL Number MFCD00143949