Learn More
Chlorendic anhydride, 95%, may cont. up to 3% chlorendic acid, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0734918
Description
Chlorendic anhydride is used in the synthesis of polymers, used in production of fiberglass-reinforced resins for chemical industry equipment. It can be used to make alkyd resins for use in special inks and paints. It is used as a hardening agent in epoxy resins used in manufacture of printed circuit boards
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChlorendic anhydride is used in the synthesis of polymers, used in production of fiberglass-reinforced resins for chemical industry equipment. It can be used to make alkyd resins for use in special inks and paints. It is used as a hardening agent in epoxy resins used in manufacture of printed circuit boards
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Chlorendic anhydride | |
115-27-5 | |
50 g | |
C9H2Cl6O3 | |
92693 | |
chlorendic anhydride, het anhydride, chloran 542, 4,7-methanoisobenzofuran-1,3-dione, 4,5,6,7,8,8-hexachloro-3a,4,7,7a-tetrahydro, 1,4,5,6,7,7-hexachloro-5-norbornene-2,3-dicarboxylic anhydride, chlorendic anhydride van, 4,5,6,7,8,8-hexachloro-3a,4,7,7a-tetrahydro-4,7-methanoisobenzofuran-1,3-dione, hexachloroendomethylene tetrahydrophthalic anhydride, hexachloro-5-norbornene-2,3-dicarboxylic anhydride, 5-norbornene-2,3-dicarboxylic anhydride, 1,4,5,6,7,7-hexachloro | |
FLBJFXNAEMSXGL-UHFFFAOYSA-N | |
370.812 | |
370.83 |
235°C to 239°C | |
1.73 | |
may cont. up to 3% chlorendic acid | |
MFCD00080438 | |
14,2086 | |
Insoluble in water. | |
C12C(C(=O)OC1=O)C3(C(=C(C2(C3(Cl)Cl)Cl)Cl)Cl)Cl | |
8265 | |
96% |
Chemical Identifiers
115-27-5 | |
370.812 | |
FLBJFXNAEMSXGL-UHFFFAOYSA-N | |
8265 |
C9H2Cl6O3 | |
MFCD00080438 | |
chlorendic anhydride, het anhydride, chloran 542, 4,7-methanoisobenzofuran-1,3-dione, 4,5,6,7,8,8-hexachloro-3a,4,7,7a-tetrahydro, 1,4,5,6,7,7-hexachloro-5-norbornene-2,3-dicarboxylic anhydride, chlorendic anhydride van, 4,5,6,7,8,8-hexachloro-3a,4,7,7a-tetrahydro-4,7-methanoisobenzofuran-1,3-dione, hexachloroendomethylene tetrahydrophthalic anhydride, hexachloro-5-norbornene-2,3-dicarboxylic anhydride, 5-norbornene-2,3-dicarboxylic anhydride, 1,4,5,6,7,7-hexachloro | |
C12C(C(=O)OC1=O)C3(C(=C(C2(C3(Cl)Cl)Cl)Cl)Cl)Cl |
Safety and Handling
GHS H Statement
H350-H315-H319-H335
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H335-H350
EINECSNumber : 204-077-3
RTECSNumber : RB9080000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only