Please login to your online account to display your discounted pricing

Alfa Aesar™ Chloranilic acid mercury(II) salt, 99%

Chemical Identifiers

Chemical Name or Material Chloranilic acid mercury(II) salt
CAS 33770-60-4
Molecular Formula C6Cl2HgO4
Formula Weight 407.56
MDL Number MFCD00044407
UN Number UN2025
Synonym 2,5-Dichloro-3,6-dihydroxy-1,4-benzoquinone mercury(II) salt; Mercury(II) chloranilate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1217306

  SDS
alfa aesar™
L1217306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 407.56
Chemical Name or Material Chloranilic acid mercury(II) salt
Molecular Formula C6Cl2HgO4
MDL Number MFCD00044407
CAS 33770-60-4
TSCA TSCA
Assay 99%
UN Number UN2025
Synonym 2,5-Dichloro-3,6-dihydroxy-1,4-benzoquinone mercury(II) salt; Mercury(II) chloranilate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.
RTECS Number OW2300000
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.