Please login to your online account to display your discounted pricing

Chloranilic acid, 99+%, ACROS Organics™

$119.39 - $119.39

Chemical Identifiers

Chemical Name or Material Chloranilic acid
CAS 87-88-7
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C6H2Cl2O4
Formula Weight 208.99
MDL Number MFCD00001596
Synonym 2,5-Dichloro-3,6-dihydroxy-1,4-benzoquinone
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC108482500

  SDS
thermo fisher scientific
AC108482500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC108481000

  SDS
thermo fisher scientific
108481000
Glass bottle 100g Each for $119.39
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 208.99
Physical Form Fine Powder
Quantity 250g
Color Orange to red
Beilstein 08, 379
Merck Index 15, 2079
Sulfated Ash 0.1% max.
Chemical Name or Material Chloranilic acid
Molecular Formula C6H2Cl2O4
CAS 87-88-7
Assay Percent Range 99+%
MDL Number MFCD00001596
Packaging Glass bottle
Synonym 2,5-Dichloro-3,6-dihydroxy-1,4-benzoquinone
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.