Please login to your online account to display your discounted pricing

Chloranilic acid, 99+%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  108481000

 View more versions of this product

Catalog No. AC108481000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 208.99
Physical Form Fine Powder
Assay ≥99.0%
Quantity 100g
Color Orange to red
Merck Index 15, 2079
Beilstein 08, 379
Infrared Spectrum Authentic
Synonym 2,5-Dichloro-3,6-dihydroxy-1,4-benzoquinone
Packaging Glass bottle
Sulfated Ash 0.1% max.
Chemical Name or Material Chloranilic acid
CAS 87-88-7
Assay Percent Range 99+%
Molecular Formula C6H2Cl2O4
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
MDL Number MFCD00001596
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Chloranilic acid
CAS 87-88-7
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C6H2Cl2O4
Formula Weight 208.99
MDL Number MFCD00001596
Synonym 2,5-Dichloro-3,6-dihydroxy-1,4-benzoquinone
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents