Please login to your online account to display your discounted pricing

Alfa Aesar™ Chloramphenicol, 10mg/mL in ethanol, sterile-filtered

$45.26 - $45.26

Chemical Identifiers

Chemical Name or Material Chloramphenicol
CAS 56-75-7
Name Note 10mg/mL in ethanol, sterile-filtered
MDL Number MFCD00078159
UN Number UN1987
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67273AB

 
alfa aesar™
J67273AB
10mL Each for $45.26

AAJ67273AC

 
alfa aesar™
J67273AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Chloramphenicol
MDL Number MFCD00078159
TSCA TSCA
Physical Form Liquid
CAS 56-75-7
UN Number UN1987
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H350-H361-H371
Highly flammable liquid and vapor.
May cause cancer.
Suspected of damaging fertility or the unborn child.
May cause damage to organs.
Health Hazard 3 GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 -30° to -10°C
Quantity 10mL
Name Note 10mg/mL in ethanol, sterile-filtered
DOT Information Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
RTECS Number AB6825000
Safety and Handling

Danger

GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H350-H361-H371
Highly flammable liquid and vapor.
May cause cancer.
Suspected of damaging fertility or the unborn child.
May cause damage to organs.

-30° to -10°C