Please login to your online account to display your discounted pricing

α-Chloralose 97%, ACROS Organics™

$27.00 - $239.58

Chemical Identifiers

Chemical Name or Material α-Chloralose
Name Note 97%
CAS 15879-93-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H11Cl3O6
Formula Weight 309.52
MDL Number MFCD00005542
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC147790250

  SDS
thermo fisher scientific
147790250
25g Each for $27.00

AC147791000

  SDS
thermo fisher scientific
147791000
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC147795000

  SDS
thermo fisher scientific
147795000
500g Each for $239.58
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 309.52
Quantity 25g
Color White to very slightly yellow
EINECS Number 240-016-7
TSCA TSCA
RTECS Number FM9450000
Melting Point 176° to 182°C
Merck Index 15, 2072
CAS Min % 96.0
CAS Max % 100.0
Residue after Ignition 0.3% max.
Molecular Formula C8H11Cl3O6
MDL Number MFCD00005542
CAS 15879-93-3
Chemical Name or Material α-Chloralose
Packaging Glass bottle
Name Note 97%
Specific Rotation +16° to +20° (20°C, 589nm) (c=2, C2H5OH)
Assay Percent Range 97%
Infrared Spectrum Authentic
Synonym 1,2-O-(2,2,2-Trichloroethylidene)-α-D-glucofuranose
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.