Please login to your online account to display your discounted pricing

Chlor. Herbicides Mixt. (Acids or Methyl Esters) (For EPA 8150, 8151)


Quantity 1x1mL (Methyl Esters)
Packaging Ampule

Manufacturer: ultra scientific  HBM815M1

 View more versions of this product

Catalog No. US-HBM-815M-1

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1x1mL (Methyl Esters)
Methods For EPA Methods 8150, 8151
Chemical Name or Material Chlorinated Herbicides Mixtures (Acids or Methyl Esters)
Concentration or Composition (by Analyte or Components) Each component at 100μg/mL in Methanol:2,4-DSilvex™2,4,5-T
Packaging Ampule

Specifications

Chemical Name or Material Chlorinated Herbicides Mixtures (Acids or Methyl Esters)