Please login to your online account to display your discounted pricing

Chlor. Herbicides Mixt. (Acids or Methyl Esters) (For EPA 8150, 8151)


Quantity 1x1mL (Acids)
Packaging Ampule

Manufacturer: ultra scientific  HBM815A1

 View more versions of this product

Catalog No. US-HBM-815A-1

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1x1mL (Acids)
Methods For EPA Methods 8150, 8151
Concentration or Composition (by Analyte or Components) Each component at 100μg/mL in Methanol:2,4-DSilvex™2,4,5-T
Packaging Ampule
Chemical Name or Material Chlorinated Herbicides Mixtures (Acids or Methyl Esters)

Specifications

Chemical Name or Material Chlorinated Herbicides Mixtures (Acids or Methyl Esters)