Chlor. Herbicides Mixt. (Acids or Methyl Esters) (For EPA 8150, 8151)
$82.09 - $87.34
Chemical Identifiers
Chemical Name or Material Chlorinated Herbicides Mixtures (Acids or Methyl Esters)
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
US-HBM-815A-1 Ultra Scientific
HBM815A1
Ampule 1x1mL (Acids) Each for $82.09
US-HBM-815M-1 Ultra Scientific
HBM815M1
Ampule 1x1mL (Methyl Esters) Each for $87.34
Specifications Description & Specifications
Chemical Identifiers
Chlorinated Herbicides Mixtures (Acids or Methyl Esters)
Specifications
1x1mL (Acids)
Each component at 100μg/mL in Methanol:2,4-DSilvex™2,4,5-T
Chlorinated Herbicides Mixtures (Acids or Methyl Esters)
For EPA Methods 8150, 8151
Ampule