Please login to your online account to display your discounted pricing

Chlor. Herbicides Mixt. (Acids or Methyl Esters) (For EPA 615, 8150B, App. IX)


Quantity 1x1mL (Acids)
Packaging Ampule

Manufacturer: ultra scientific  HBM8150A1

 View more versions of this product

Catalog No. US-HBM-8150A1

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1x1mL (Acids)
Concentration or Composition (by Analyte or Components) Each component in Methanol:
2,4-D    •100μg/mL
Dalapon™    •250μg/mL
2,4-DB    •100μg/mL
Dicamba    •10μg/mL
Dichloroprop    •100μg/mL
Dinoseb™    •50μg/mL
MCPA    •10,000μg/mL
MCPP    •10,000μg/mL
Silvex™(2,4,5-TP)    •10μg/mL
2,4,5-T    •10μg/mL
Methods For EPA Methods 615, 8150B, Appendix IX
Chemical Name or Material Chlorinated Herbicides Mixtures (Acids or Methyl Esters)
Packaging Ampule

Specifications

Chemical Name or Material Chlorinated Herbicides Mixtures (Acids or Methyl Esters)