Please login to your online account to display your discounted pricing

Chlor. Herbicides Mixt. (Acids or Methyl Esters) (For EPA 615, 8150B, App. IX)

$250.43 - $263.49

Chemical Identifiers

Chemical Name or Material Chlorinated Herbicides Mixtures (Acids or Methyl Esters)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

US-HBM-8150A1

 
ultra scientific
HBM8150A1
Ampule 1x1mL (Acids) Each for $250.43

US-HBM-8150M1

 
ultra scientific
HBM8150M1
Ampule 1x1mL (Methyl Esters) Each for $263.49
Description & Specifications

Specifications

Quantity 1x1mL (Acids)
Concentration or Composition (by Analyte or Components) Each component in Methanol:
2,4-D    •100μg/mL
Dalapon™    •250μg/mL
2,4-DB    •100μg/mL
Dicamba    •10μg/mL
Dichloroprop    •100μg/mL
Dinoseb™    •50μg/mL
MCPA    •10,000μg/mL
MCPP    •10,000μg/mL
Silvex™(2,4,5-TP)    •10μg/mL
2,4,5-T    •10μg/mL
Methods For EPA Methods 615, 8150B, Appendix IX
Chemical Name or Material Chlorinated Herbicides Mixtures (Acids or Methyl Esters)
Packaging Ampule