missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CHIR 99021 trihydrochloride, Tocris™

Hydrochloride salt of CHIR 99021; selective GSK-3 inhibitor

$295.43 - $1,222.50

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C22H18Cl2N8·3HCl
Formula Weight 574.72
Synonym 6-[[2-[[4-(2, 4-Dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)-2-pyrimidinyl]amino]ethyl]amino]-3-pyridinecarbonitrile trihydrochloride
Chemical Name or Material CHIR 99021 trihydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
495310 R & D Systems
495310
10mg Each for $295.43
495350 R & D Systems
495350
50mg Each for $1,222.50
Specifications Description & Specifications

Chemical Identifiers

>99%
574.72
CHIR 99021 trihydrochloride
C22H18Cl2N8·3HCl
6-[[2-[[4-(2, 4-Dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)-2-pyrimidinyl]amino]ethyl]amino]-3-pyridinecarbonitrile trihydrochloride

Specifications

10mg
C22H18Cl2N8·3HCl
6-[[2-[[4-(2, 4-Dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)-2-pyrimidinyl]amino]ethyl]amino]-3-pyridinecarbonitrile trihydrochloride
>99%
574.72
CHIR 99021 trihydrochloride

Safety and Handling

Recommended Storage : Store at -20°C