Please login to your online account to display your discounted pricing

CHES, ULTROL™ Grade, Calbiochem™, EMD Millipore

A zwitterionic buffer that is useful in the pH 8.6 to 10.0 range

$81.70 - $81.70

Chemical Identifiers

Assay Percent Range ≥99.0%
Formula Weight 207.3
CAS 103-47-9
Chemical Name or Material CHES
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

239779100GM

 
emd millipore corporation
239779100GM
Each for $81.70
Description & Specifications

Specifications

Formula Weight 207.3
Quantity 100g
Chemical Name or Material CHES
Purity Grade Notes by titration
Assay Percent Range ≥99.0%
CAS 103-47-9
Synonym 2-(N-Cyclohexylamino)ethanesulfonic Acid
Solubility Information Easily soluble in cold water
Recommended Storage 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
Safety and Handling

15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).