missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ CHES, 0.5M buffer soln, pH 10.0

$84.96 - $133.08

Chemical Identifiers

CAS 103-47-9
Molecular Formula C8H17NO3S
Molecular Weight (g/mol) 207.288
MDL Number MFCD00003835
InChI Key MKWKNSIESPFAQN-UHFFFAOYSA-N
PubChem CID 66898
IUPAC Name 2-(cyclohexylamino)ethanesulfonic acid
SMILES C1CCC(CC1)NCCS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63419AE View Documents Alfa Aesar™
J63419AE
100mL Each for $84.96
 
AAJ63419AK View Documents Alfa Aesar™
J63419AK
250mL Each for $133.08
 
Specifications

Chemical Identifiers

103-47-9
207.288
MKWKNSIESPFAQN-UHFFFAOYSA-N
2-(cyclohexylamino)ethanesulfonic acid
C8H17NO3S
MFCD00003835
66898
C1CCC(CC1)NCCS(=O)(=O)O

Specifications

103-47-9
MFCD00003835
MKWKNSIESPFAQN-UHFFFAOYSA-N
2-(cyclohexylamino)ethanesulfonic acid
66898
10.0
CHES buffer soln.
C8H17NO3S
0.5M buffer soln
C1CCC(CC1)NCCS(=O)(=O)O
207.288
Liquid
100mL
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

TSCA : TSCA