missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chenodeoxycholic acid

Bile acid that induces apoptosis through protein kinase C signaling pathways

$105.49 - $323.85

Chemical Identifiers

CAS 474-25-9
Molecular Formula C24H39O4-
Molecular Weight (g/mol) 391.572
MDL Number MFCD00064142
InChI Key RUDATBOHQWOJDD-BSWAIDMHSA-M
Synonym chenodexycholic acid, chenodeoxycholate anion, chenodeoxycholate 1-, 3alpha, 7alpha-dihydroxy-5beta-cholanic acid, 3alpha,7alpha-dihydroxy-5beta-cholan-24-oate, 3alpha,7alpha-dihydroxy-5beta-cholan-24-oic acid anion, 4r-4-3r,5s,7r,8r,9s,10s,13r,14s,17r-10,13-dimethyl-3,7-bis oxidanyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1h-cyclopenta a phenanthren-17-yl pentanoate, 4r-4-3r,5s,7r,8r,9s,10s,13r,14s,17r-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1h-cyclopenta a phenanthren-17-yl pentanoate
PubChem CID 9548590
ChEBI CHEBI:36234
IUPAC Name (4R)-4-[(3R,5S,7R,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
SMILES CC(CCC(=O)[O-])C1CCC2C1(CCC3C2C(CC4C3(CCC(C4)O)C)O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6036406 View Documents Alfa Aesar™
J6036406
5g Each for $105.49
 
AAJ6036414 View Documents Alfa Aesar™
J6036414
25g Each for $323.85
 
Description

Description

Solubility
Soluble in water, dimethyl sulfoxide, ethanol and dimethylformamide.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

474-25-9
391.572
RUDATBOHQWOJDD-BSWAIDMHSA-M
9548590
(4R)-4-[(3R,5S,7R,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
C24H39O4-
MFCD00064142
chenodexycholic acid, chenodeoxycholate anion, chenodeoxycholate 1-, 3alpha, 7alpha-dihydroxy-5beta-cholanic acid, 3alpha,7alpha-dihydroxy-5beta-cholan-24-oate, 3alpha,7alpha-dihydroxy-5beta-cholan-24-oic acid anion, 4r-4-3r,5s,7r,8r,9s,10s,13r,14s,17r-10,13-dimethyl-3,7-bis oxidanyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1h-cyclopenta a phenanthren-17-yl pentanoate, 4r-4-3r,5s,7r,8r,9s,10s,13r,14s,17r-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1h-cyclopenta a phenanthren-17-yl pentanoate
CHEBI:36234
CC(CCC(=O)[O-])C1CCC2C1(CCC3C2C(CC4C3(CCC(C4)O)C)O)C

Specifications

+12° (c=2 in Ethanol)
164°C to 168°C
474-25-9
MFCD00064142
14,2054
RUDATBOHQWOJDD-BSWAIDMHSA-M
(4R)-4-[(3R,5S,7R,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
9548590
392.57
Chenodeoxycholic acid
White
5g
C24H39O4-
3219887
chenodexycholic acid, chenodeoxycholate anion, chenodeoxycholate 1-, 3alpha, 7alpha-dihydroxy-5beta-cholanic acid, 3alpha,7alpha-dihydroxy-5beta-cholan-24-oate, 3alpha,7alpha-dihydroxy-5beta-cholan-24-oic acid anion, 4r-4-3r,5s,7r,8r,9s,10s,13r,14s,17r-10,13-dimethyl-3,7-bis oxidanyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1h-cyclopenta a phenanthren-17-yl pentanoate, 4r-4-3r,5s,7r,8r,9s,10s,13r,14s,17r-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1h-cyclopenta a phenanthren-17-yl pentanoate
CC(CCC(=O)[O-])C1CCC2C1(CCC3C2C(CC4C3(CCC(C4)O)C)O)C
391.572
CHEBI:36234
Needle Crystals
Safety and Handling

Safety and Handling

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : FZ1980000