Please login to your online account to display your discounted pricing

Chemistry Bulletin Board Set

A colorful reference bulletin board set.

$19.60 - $19.60

Specifications

Category Chemistry
Format Set
For Use With (Application) Colorful reference bulletin board set for students
Grade 4 to 8
View More Specs
Products
Catalog Number Mfr. No. Includes Product Type Price Quantity    

S02424

 
fisher science education
CD-1938
4 Charts (24 x 17 in.), resource guide Chemistry Bulletin Board Set Each for $19.60
Description & Specifications

Specifications

Category Chemistry
Format Set
For Use With (Application) Colorful reference bulletin board set for students
Grade 4 to 8

Includes:

  • One periodic table (48 × 34 in. when assembled)
  • A resource guide