Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves.
$4,764.12 - $4,764.12
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products ${productFamilyLength}
Catalog Number Mfr. No. Actuation Ports Stator Material Price Quantity  
Catalog Number Mfr. No. Actuation Ports Stator Material Price Quantity  
03-052-404 Thermo Scientific™
C21000EP
Microelectric 10 N60 stainless Each for $4,764.12
Specifications
Description
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves. It can be actuated manually or with a microelectronic actuator. With 1/16 in. fittings and 0.010 in. ports, it is available in six-port or ten-port configurations and includes a 5μL sample loop.
  • 1/16″ fittings
  • 0.010″ ports
  • Available in 6-port or 10-port configurations
  • Available with manual or microelectric actuator