Learn More
MilliporeSigma™ Chemicon™ Millicoat™ Human Collagen Type IV Coated Strips (96-Wells)
Used for cell adhesion studies
Supplier: MilliporeSigma™ ECM105
Description
- Provided as 12 x 8-well removable strips in a plate frame for convenience and flexibility in designing assays
- Wells in rows A to G have been coated with Human Collagen Type IV
- Row H of each strip is coated with BSA which serves as negative assay control
Specifications
Enzyme Assay | |
1 x 96-well Plate | |
Collagen, Human Type IV Coated Strip | |
May be stored at 2° to 8°C in the foil pouch for at least 3 months |