Please login to your online account to display your discounted pricing

EMD Millipore™ Chemicon™ Laminin, Mouse Purified


For Cell Culture applications

Manufacturer: emd millipore™  CC0955MG

 View more versions of this product

Catalog No. CC0955MG

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
For Use With (Application) Cell culture
Format Liquid
Formulation Liquid; in 0.05 M Tris-HCl, pH 7.4, containing 0.15 M NaCl. 
Purity >95% by SDS-PAGE 
Storage Requirements Store at -20°C for up to six months. 
Quantity 5mg
Source Isolated from Engelbreth-Holm-Swarm (EHS) mouse sarcoma
Type Laminin, Mouse Purified