Please login to your online account to display your discounted pricing

EMD Millipore™ Chemicon™ Human Plasma Fibronectin Purified Protein


Suitable for cell propagation in vitro

Manufacturer: emd millipore™  FC010100MG

 View more versions of this product

Catalog No. FC010100MG

  • / Each
 

Description & Specifications

Specifications

Format Liquid
Concentration 1mg/mL
Quantity 100mg
Storage Requirements Maintain at 2° to 8°C for up to 6 months from date of receipt. Do not freeze.
Type Human plasma fibronectin purified protein
For Use With (Application) Cell culture
pH 7.5
Formulation Liquid in 150 mM NaCl, 10 mM sodium phosphate, pH 7.5, sterile filtered, containing no preservatives.
Source Human donor plasma