missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Human Collagen Type IV

Purified by using differential salt precipitation

Supplier:  MilliporeSigma™ CC076

Catalog No. CC076


Explore available promotions

Description

Description

  • Major structural element of basal membranes
  • Native triple helix
  • Extracted from washed dissected tissue into dilute acetic acid after mild pepsin treatment
  • Purity and retention controlled by SDS-PAGE and by reaction with anti-collagen type specific monoclonal antibodies
  • Molecular weight: 300 kd
  • Solubility: 10mg/mL
Specifications

Specifications

1 mg/mL
100μL of 0.5M acetic acid solution, pH 2.5. Salt-free, no preservative
Human placenta, negative for HBsAg and HIV antibodies
Store at -20°C for up to 12 months. Avoid repeated freeze/thaw.
95% by SDS-PAGE
Cell Culture
100 μg
Collagen, Human Type IV
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions