Please login to your online account to display your discounted pricing

EMD Millipore™ Chemicon™ Fibronectin α Chymotryptic Fragment (Heparin-binding region), Human Purified


For Cell Culture applications

Manufacturer: emd millipore™  F1903

 View more versions of this product

Catalog No. F1903

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
For Use With (Application) Cell culture
Format  Lyophilized in sodium citrate buffer
Storage Requirements Maintain material at -20°C for 3 months or -70°C for six months.
Quantity 500μg
Source Human plasma cryoprecipitate (plasma fibronectin), assayed negative for hepatitis B virus, hepatitis C Virus (HCV), HIV-1, HIV-2, HTLV-1, HTLV-2 and Treponema pallidum.
Type Fibronectin 40 kd α Chymotryptic Fragment
pH 7.4