Please login to your online account to display your discounted pricing

Chemical Demonstrations


Manufacturer: fisher science education  0299128601

 View more versions of this product

Catalog No. S83138

  • / Each
 

Description & Specifications

Specifications

Includes Lists of references
Product Type Chemical Demonstrations Volume 4
Category Chemistry
For Use With (Application) Demonstrate to teach science to students
Format Hardcover Book