Please login to your online account to display your discounted pricing

Chemical Demonstrations

$60.00 - $199.95

Specifications

Category Chemistry
Format Book
For Use With (Application) Demonstrate to teach science to students
Includes Lists of references
View More Specs
Includes:  There are lists of references included.
The discussion section provides information about the chemical phenomena and the principles illustrated by each demonstration. Over 275 demonstrations covering 11 different topics are in these volumes. Hard cover.
Products
Catalog Number Mfr. No. Product Type Price Quantity    

S83137

 
fisher science education
0299119508
Chemical Demonstrations Volume 3 Each for $60.30

S83138

 
fisher science education
0299128601
Chemical Demonstrations Volume 4 Each for $60.40

S83135

 
fisher science education
0299088901
Chemical Demonstrations Volume 1 Each for $60.00

S83136

 
fisher science education
0299101304
Chemical Demonstrations Volume 2 Each for $60.00

S99025

 
fisher science education
FISHERMAKE
Chemical Demonstrations book Each for $199.95
Description & Specifications

Specifications

Category Chemistry
Format Book
For Use With (Application) Demonstrate to teach science to students
Includes Lists of references

Use demonstrations to teach science to your students

Bassam Shakhashiri, internationally known chemist, lecture-demonstrator, educator, and teacher of teachers has compiled four books of demonstrations. Since the first volume appeared in 1983, he has been urged by teachers to continue making these demonstrations available. Volume 4 was completed in 1992. It contains a cumulative index for Volumes 1 to 4.
Each volume contains demonstration overviews, materials lists, step-by-step instructions for preparation and presentation of the demonstrations, and sections about hazards, disposal, and discussion.