Please login to your online account to display your discounted pricing

Chemical Demonstrations


Manufacturer: fisher science education  FISHERMAKE

 View more versions of this product

Catalog No. S99025

  • / Each
 

Description & Specifications

Specifications

Category Chemistry
Includes Lists of references
Format Book
For Use With (Application) Demonstrate to teach science to students
Product Type Chemical Demonstrations book