Chemglass Distilling Bulbs
$94.46 - $114.51
Specifications
Product Type Distilling Bulb Kugelrohr
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Diameter (Metric) Outer Height (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity Diameter (Metric) Outer Height (Metric) Price Quantity  
CG122710 Chemglass Inc
CG122710
10mL 30mm 85mm Each for $94.46
CG122711 Chemglass Inc
CG122711
15mL 35mm 90mm Each for $97.97
CG122713 Chemglass Inc
CG122713
50mL 50mm 105mm Each for $114.51
CG122712 Chemglass Inc
CG122712
25mL 40mm 98mm Each for $105.86
Specifications Description & Specifications
Specifications
Distilling Bulb Kugelrohr