Please login to your online account to display your discounted pricing

CHEMetrics™ COD Vials Kits

$461.86 - $461.86

Specifications

Composition >64% Sulfuric Acid, <34% Deionized Water, <1% Mercuric Sulfate, <1% Silver Sulfate, <0.5% Potassium Dichromate
For Use With (Application) Determination of the oxidizable organic matter content of wastewater
For Use With (Equipment) Digestor block as well as a photometer or spectrophotometer
Test Method Dichromate reactor digestion
View More Specs
Requires:  Requires the use of a Digestor Block and the V-2000 Photometer, a COD Photometer, or a spectrophotometer capable of accepting a 16mm round cell. Instruments sold separately.

 Alert

Product contains mercury - dispose according to local, state, or federal laws.

Products
Catalog Number Mfr. No. Includes Range Price Quantity    

13-299-230

 
chemetrics™
CHIK7365**
150 vials and instruction book 0-1500ppm Pack of 150 Each for $461.86
Description & Specifications

Specifications

Composition >64% Sulfuric Acid, <34% Deionized Water, <1% Mercuric Sulfate, <1% Silver Sulfate, <0.5% Potassium Dichromate
For Use With (Application) Determination of the oxidizable organic matter content of wastewater
For Use With (Equipment) Digestor block as well as a photometer or spectrophotometer
Test Method Dichromate reactor digestion

Low range vials

  • Contains everything needed (except distilled water) to perform 149 tests
  • Includes instruction book
  • Comes in cardboard box
  • 2-hour digestion time
  • Dichromate reactor digestion

A fresh reagent ampoule blank must be prepared for each series of tests; therefore the number of samples that can be tested with each kit will vary.

Low range vials EPA approved.