Please login to your online account to display your discounted pricing

CHEMetrics™ COD Vials Kits


Manufacturer: chemetrics™  CHIK7365**

 View more versions of this product

Requires the use of a Digestor Block and the V-2000 Photometer, a COD Photometer, or a spectrophotometer capable of accepting a 16mm round cell. Instruments sold separately.

 Alert

Product contains mercury - dispose according to local, state, or federal laws.

Catalog No. 13-299-230

  • / Pack of 150 Each
 

Description & Specifications

Specifications

Certifications/Compliance USEPA
Composition >64% Sulfuric Acid, <34% Deionized Water, <1% Mercuric Sulfate, <1% Silver Sulfate, <0.5% Potassium Dichromate
Range 0-1500ppm
For Use With (Application) Determination of the oxidizable organic matter content of wastewater
For Use With (Equipment) Digestor block as well as a photometer or spectrophotometer
Includes 150 vials and instruction book
Test Method Dichromate reactor digestion