CHEMetrics™ COD Reagent Vials
Contain premeasured reagents for accurate, economical determination of COD
Manufacturer:  CHEMetrics™ CHIK7361S
 View more versions of this product
Catalog No. 13-299-733
Specifications
Description
COD Testing, Mercury-Free Dichromate Reactor Digestion Method, 25 PTFE-Lined Screw Cap Vials, 0-1500ppm, 30ppm MDL
  • No need to modify or recalibrate existing equipment or procedures
  • Optical-quality glass for accuracy
  • Available in mercury and mercury-free versions
  • Screw caps lined with PTFE for inertness
  • Standard 5/8 in. (16mm) diameter for compatibility with standard COD reactors and spectrophotometers; can also be used in micro-COD reactors
  • Follow the EPA-approved method for compliance reporting and use potassium dichromate as the oxidizing agent, silver sulfate as the catalyst, and mercury chloride to remove chloride interference
  • Mercury-free version uses the same oxidizing agent and catalyst without the chloride compensator
Description & Specifications
Specifications
>64% Sulfuric Acid, <34% Deionized Water, <1% Mercuric Sulfate, <1% Silver Sulfate, <0.5% Potassium Dichromate
Premeasured reagents for accurate, economical determination of COD, determination of the oxidizable organic matter content of wastewater
PTFE
Mercury-free
0 to 1500ppm
Instruction book
25.0