Please login to your online account to display your discounted pricing
CHEMetrics™ COD Reagent Vials
Contain premeasured reagents for accurate, economical determination of COD
Manufacturer: chemetrics™  CHIK7351S
 View more versions of this product
Catalog No. 13-299-731
  • / Pack of 25 Each
Description & Specifications
Specifications
Composition >64% Sulfuric Acid, <34% Deionized Water, <1% Mercuric Sulfate, <1% Silver Sulfate, <0.5% Potassium Dichromate
Test Method Mercury-free
For Use With (Application) Premeasured reagents for accurate, economical determination of COD, determination of the oxidizable organic matter content of wastewater
Liner Material PTFE
Range 0 to 150ppm
No. of Tests 25.0
Includes Instruction book