CHEMetrics™ Chemets™ Water Testing Kits
Provide results in two minutes or less
Manufacturer:  CHEMetrics™ CHIK4502
 View more versions of this product
Catalog No. 13-299-216C
Specifications Description & Specifications
Specifications
Chemets™ Water Testing Kit
Water testing kits for chlorine (DPD), copper, dissolved oxygen, hydrogen peroxide, phenols, phosphate, silica, sulfide and more
30.0
2 to 20ppm as CaCO3
Cardboard box, 30 ampoules of reagent with valve assemblies, titrettor, 25mL sample cup, instructions
Total EDTA