CHEMetrics CHEMets Filming Amine Visual Water Test Kit
Provides results in two minutes or less
Supplier: CHEMetrics K1001
Includes: REMOVE
Description
Provide results in two minutes or less
CHEMets Filming Amine kit (K-1001) comes in a cardboard box and contains everything needed to perform 20 tests: Refill, Comparator, reaction tube with lid, tip breaking tool and instructionsSpecifications
Filming Amines | |
20 | |
Methyl Orange |
Cardboard box, Refill, Comparator, reaction tube with lid, tip breaking tool and instructions | |
0 to 1 ppm | |
CHEMets Visual Water Testing Kit |
Warning! Prop 65