Please login to your online account to display your discounted pricing
CHEMetrics™ Chemets™ Water Testing Kits
Provide results in two minutes or less
Manufacturer: chemetrics™  CHIK2500
 View more versions of this product
Catalog No. 13-299-248
  • / Pack
Description & Specifications
Specifications
No. of Tests 30.0
For Use With (Application) Water testing kits for chlorine (DPD), copper, dissolved oxygen, hydrogen peroxide, phenols, phosphate, silica, sulfide and more
Includes Plastic case, 30 ampoules of reagent, comparator(s), snap cup (holds water sample), instructions
Method DPD
Analytes Free and Total Chlorine
Product Type Chemets™ Water Testing Kit
Range 0.1 to 2ppm