Please login to your online account to display your discounted pricing

CHEMetrics™ Chemets™ Test Kit Refills


Provide results in two minutes or less

Manufacturer: chemetrics™  CHIR9010

 View more versions of this product

Catalog No. 13-299-508L

  • / Pack of 30 Each
 

Description & Specifications

Specifications

Analytes Silica
Product Type Chemets™ Refill
Range 0 to 1, 1 to 10ppm
For Use With (Equipment) Silica testing kits
Includes 30 ampoules
Shelf Life 11 Months
Method Heteropoly Blue
Accessories

11 Months