Please login to your online account to display your discounted pricing

CHEMetrics™ Chemets™ Test Kit Refills


Provide results in two minutes or less

Manufacturer: chemetrics™  CHIR6902

 View more versions of this product

Catalog No. 13-299-508F

  • / Pack of 30 Each
 

Description & Specifications

Specifications

Method Cadmium Reduction
Shelf Life 2 Years
Includes 30 ampoules
Range 0 to 3ppm
For Use With (Equipment) Nitrate testing kits
Test Time 15 min.
Analytes Cadmium
Product Type Chemets™ Refill
Accessories

2 Years