CHEMetrics CHEMets Nitrite Test Kit Refill
Provides results in two minutes or less
Supplier: CHEMetrics R7002
Description
- Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
- To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
- In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
- Comparators have two-year shelf life
Specifications
Nitrite | |
30 self-filling CHEMets ampoules to refill kit K-7004 | |
0.2 to 2 ppm | |
10 min. |
C-7004 comparators | |
30 | |
Azo dye formation | |
CHEMets Refill |
Warning! Prop 65