Please login to your online account to display your discounted pricing

CHEMetrics™ Chemets™ Test Kit Refills


Provide results in two minutes or less

Manufacturer: chemetrics™  CHIR7512

 View more versions of this product

Catalog No. 13-299-200B

  • / Pack of 30 Each
 

Description & Specifications

Specifications

Method Indigo Carmine
Shelf Life 2 Years
Includes 30 ampoules
Test Time 2 min.
Product Type Chemets™ Refill
For Use With (Equipment) Oxygen (dissolved) testing kits
Analytes Dissolved Oxygen
Range 1 to 12ppm
Accessories

2 Years