missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chelidonic Acid Monohydrate, TCI America™

$165.00 - $548.48

Chemical Identifiers

CAS 6003-94-7
Molecular Formula C7H6O7
Molecular Weight (g/mol) 202.118
MDL Number MFCD00149433
InChI Key KJZJAKSEIQOKAK-UHFFFAOYSA-N
Synonym chelidonic acid monohydrate, compound xi* hydrate, 4-oxo-4h-pyran-2,6-dicarboxylic acid monohydrate, chelidonic acidmonohydrate, chelidonic acid 1-hydrate, c7h4o6.h2o, 2,6-dicarboxy-4-oxo-4h-pyran monohydrate, 4-oxo-4h-pyran-2,6-dicarboxylic acid hydrate, 4h-pyran-2,6-dicarboxylicacid, 4-oxo-, hydrate 1:1
PubChem CID 51346158
IUPAC Name 4-oxopyran-2,6-dicarboxylic acid;hydrate
SMILES C1=C(OC(=CC1=O)C(=O)O)C(=O)O.O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C09555G View Documents TCI America
C09555G
5g Each for $165.00
 
C095525G View Documents TCI America
C095525G
25g Each for $548.48
 
Specifications

Chemical Identifiers

6003-94-7
202.118
KJZJAKSEIQOKAK-UHFFFAOYSA-N
51346158
C1=C(OC(=CC1=O)C(=O)O)C(=O)O.O
C7H6O7
MFCD00149433
chelidonic acid monohydrate, compound xi* hydrate, 4-oxo-4h-pyran-2,6-dicarboxylic acid monohydrate, chelidonic acidmonohydrate, chelidonic acid 1-hydrate, c7h4o6.h2o, 2,6-dicarboxy-4-oxo-4h-pyran monohydrate, 4-oxo-4h-pyran-2,6-dicarboxylic acid hydrate, 4h-pyran-2,6-dicarboxylicacid, 4-oxo-, hydrate 1:1
4-oxopyran-2,6-dicarboxylic acid;hydrate

Specifications

Yellow
6003-94-7
MFCD00149433
KJZJAKSEIQOKAK-UHFFFAOYSA-N
4-oxopyran-2,6-dicarboxylic acid;hydrate
51346158
Crystal-Powder at 20°C
Chelidonic Acid Monohydrate
5g
C7H6O7
chelidonic acid monohydrate, compound xi* hydrate, 4-oxo-4h-pyran-2,6-dicarboxylic acid monohydrate, chelidonic acidmonohydrate, chelidonic acid 1-hydrate, c7h4o6.h2o, 2,6-dicarboxy-4-oxo-4h-pyran monohydrate, 4-oxo-4h-pyran-2,6-dicarboxylic acid hydrate, 4h-pyran-2,6-dicarboxylicacid, 4-oxo-, hydrate 1:1
C1=C(OC(=CC1=O)C(=O)O)C(=O)O.O
202.118
184.10
≥95.0% (T)
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1292

RTECSNumber : UP7350000

TSCA : No