missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chelidonic acid monohydrate, 96%, Thermo Scientific™

$142.11 - $538.09

Chemical Identifiers

CAS 6003-94-7
Molecular Formula C7H4O6·H2O
Molecular Weight (g/mol) 202.12
MDL Number MFCD00149433
InChI Key KJZJAKSEIQOKAK-UHFFFAOYSA-N
Synonym 2,6-dicarboxy-4-oxo-4h-pyran monohydrate, 4-oxo-4h-pyran-2,6-dicarboxylic acid hydrate, 4-oxo-4h-pyran-2,6-dicarboxylic acid monohydrate, 4h-pyran-2,6-dicarboxylicacid, 4-oxo-, hydrate 1:1, c7h4o6.h2o, chelidonic acid 1-hydrate, chelidonic acid monohydrate, chelidonic acidmonohydrate, compound xi* hydrate
PubChem CID 51346158
IUPAC Name 4-oxopyran-2,6-dicarboxylic acid;hydrate
SMILES C1=C(OC(=CC1=O)C(=O)O)C(=O)O.O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC154600050
SDS View Documents Promotion Details
Thermo Fisher Scientific
154600050
5g Glass bottle
Each for $142.11
 
AC154600250
SDS View Documents Promotion Details
Thermo Fisher Scientific
154600250
25g Plastic bottle
Each for $538.09
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

6003-94-7
202.12
KJZJAKSEIQOKAK-UHFFFAOYSA-N
51346158
C1=C(OC(=CC1=O)C(=O)O)C(=O)O.O
C7H4O6·H2O
MFCD00149433
2,6-dicarboxy-4-oxo-4h-pyran monohydrate, 4-oxo-4h-pyran-2,6-dicarboxylic acid hydrate, 4-oxo-4h-pyran-2,6-dicarboxylic acid monohydrate, 4h-pyran-2,6-dicarboxylicacid, 4-oxo-, hydrate 1:1, c7h4o6.h2o, chelidonic acid 1-hydrate, chelidonic acid monohydrate, chelidonic acidmonohydrate, compound xi* hydrate
4-oxopyran-2,6-dicarboxylic acid;hydrate

Specifications

6003-94-7
265°C
C7H4O6·H2O
MFCD00149433
18, 490
5g
KJZJAKSEIQOKAK-UHFFFAOYSA-N
4-oxopyran-2,6-dicarboxylic acid;hydrate
51346158
Powder
Chelidonic acid monohydrate, 96%
Yellow to Beige
95% min. (HPLC)
Authentic
Glass bottle
14, 2052
2,6-dicarboxy-4-oxo-4h-pyran monohydrate, 4-oxo-4h-pyran-2,6-dicarboxylic acid hydrate, 4-oxo-4h-pyran-2,6-dicarboxylic acid monohydrate, 4h-pyran-2,6-dicarboxylicacid, 4-oxo-, hydrate 1:1, c7h4o6.h2o, chelidonic acid 1-hydrate, chelidonic acid monohydrate, chelidonic acidmonohydrate, compound xi* hydrate
C1=C(OC(=CC1=O)C(=O)O)C(=O)O.O
202.12
202.12
96%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

SDS
Product Certifications

RUO – Research Use Only