Please login to your online account to display your discounted pricing

Charcoal U.S.P. Powder, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Charcoal
Name Note Activated
CAS 7440-44-0
Purity Grade USP
Physical Form Powder
Molecular Formula C
Formula Weight 12.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK4394500

 
avantor performance
439412
Glass 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 12.01
Quantity 500g
Physical Form Powder
Purity Grade USP
CAS 7440-44-0
Chemical Name or Material Charcoal
Molecular Formula C
Name Note Activated
Packaging Glass