missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ CHAPS lysis buffer

For cell lysis and protein extraction

Specifications

Chemical Name or Material CHAPS lysis buffer
MDL Number MFCD00282866
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60580AD View Documents Alfa Aesar™
J60580AD
50mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ60580AE View Documents Alfa Aesar™
J60580AE
100mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Specifications

CHAPS lysis buffer
MFCD00282866