missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ CHAPS, 98+%

$1,019.00 - $1,019.00

Chemical Identifiers

CAS 75621-03-3
Molecular Formula C32H58N2O7S
Formula Weight 614.88
MDL Number MFCD00012116
Synonym 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate
Chemical Name or Material CHAPS
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2192714 SDS Alfa Aesar™
B2192714
25g Each for $1,019.00
AAB2192703 SDS Alfa Aesar™
B2192703
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAB2192706 SDS Alfa Aesar™
B2192706
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
Buffer; zwitterionic nondenaturing detergent for solubilizing membrane proteinsCHAPS is used in the laboratory to dissolve biological macromolecules such as proteins. It acts as a non-denaturing solvent used in purification of membrane proteins. It is also useful in conjunction with nonionic detergents like Triton X-100. Further, it is used as a non-denaturing zwitterionic detergent for membrane biochemistry.

Solubility
Slightly soluble in water.

Notes
Store in a cool. Incompatible with oxidizing agent, strong acid and bases.
Description & Specifications

Chemical Identifiers

75621-03-3
614.88
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate
C32H58N2O7S
MFCD00012116
CHAPS

Specifications

75621-03-3
614.88
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate
25g
≥98%
C32H58N2O7S
MFCD00012116
156° to 158°C (dec.)
CHAPS

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning