Please login to your online account to display your discounted pricing

Alfa Aesar™ Chalcone alpha,beta-epoxide, 98%

$261.68 - $261.68

Chemical Identifiers

Chemical Name or Material Chalcone alpha,beta-epoxide
CAS 5411-12-1
Molecular Formula C15H12O2
Formula Weight 224.26
MDL Number MFCD00022336
Beilstein 164553
Synonym 2-(Benyzlidene)acetophenone epoxide; 2-Benzoyl-3-phenyloxirane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0387414

  SDS
alfa aesar™
L0387414
25g Each for $261.68
Description & Specifications

Specifications

Formula Weight 224.26
Chemical Name or Material Chalcone alpha,beta-epoxide
Molecular Formula C15H12O2
MDL Number MFCD00022336
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
CAS 5411-12-1
Synonym 2-(Benyzlidene)acetophenone epoxide; 2-Benzoyl-3-phenyloxirane
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 88° to 90°C
Quantity 25g
Beilstein 164553
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.