missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chalcone alpha,beta-epoxide, 98%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific L0387406

 View more versions of this product

Catalog No. AAL0387406



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Chalcone alpha,beta-epoxide
88°C to 90°C
MFCD00022336
1,3-diphenyl-2,3-epoxy-1-propanone, 2-benzoyl-3-phenyloxirane, ccris 2214, chalcone epoxide, chalcone oxide, methanone, phenyl 3-phenyloxiranyl, phenyl 3-phenyl-2-oxiranyl methanone, phenyl 3-phenyloxiran-2-yl methanone, trans-chalcone oxide, trans-phenyl 3-phenyloxiranyl methanone
UQGMJZQVDNZRKT-UHFFFAOYNA-N
phenyl-(3-phenyloxiran-2-yl)methanone
92219
98%
5411-12-1
C15H12O2
164553
5g
C1=CC=C(C=C1)C2C(O2)C(=O)C3=CC=CC=C3
224.26
224.26

Chemical Identifiers

5411-12-1
224.26
UQGMJZQVDNZRKT-UHFFFAOYNA-N
92219
C1=CC=C(C=C1)C2C(O2)C(=O)C3=CC=CC=C3
C15H12O2
MFCD00022336
1,3-diphenyl-2,3-epoxy-1-propanone, 2-benzoyl-3-phenyloxirane, ccris 2214, chalcone epoxide, chalcone oxide, methanone, phenyl 3-phenyloxiranyl, phenyl 3-phenyl-2-oxiranyl methanone, phenyl 3-phenyloxiran-2-yl methanone, trans-chalcone oxide, trans-phenyl 3-phenyloxiranyl methanone
phenyl-(3-phenyloxiran-2-yl)methanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 226-487-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only