missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chalcone, 97%, Acros Organics

Manufacturer:  Fisher Scientific Acros 159431000

 View more versions of this product

Catalog No. AC159431000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Chalcone
Authentic
345.0°C to 348.0°C
53.0°C to 58.0°C
97%
96% min. (GC)
C6H5CH=CHCOC6H5
07, 478
2-benzalacetophenone
DQFBYFPFKXHELB-VAWYXSNFSA-N
(E)-1,3-diphenylprop-2-en-1-one
637760
208.26
97%
>112°C
Plastic bottle
Yellow
100g
94-41-7
C15H12O
MFCD00003082
15, 2036
Solubility in water: insoluble. Other solubilities: freely soluble in ether,chloroform,benzene and,carbon disulfide,slightly soluble in alcohol
C1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2
208.26
CHEBI:48965
Granular Powder

Chemical Identifiers

94-41-7
208.26
DQFBYFPFKXHELB-VAWYXSNFSA-N
637760
(E)-1,3-diphenylprop-2-en-1-one
C15H12O
MFCD00003082
2-benzalacetophenone
CHEBI:48965
C1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2
Safety and Handling

Safety and Handling

EINECSNumber : 202-330-2

RTECSNumber : FL6900000

TSCA : TSCA

SDS Documents