Please login to your online account to display your discounted pricing

Ch 55, Tocris

Potent RAR agonist

$192.67 - $812.12

Chemical Identifiers

Chemical Name or Material Ch 55
Assay Percent Range >98%
Molecular Formula C24H28O3
Formula Weight 364.47
Synonym 4-[(1E)-3-[3, 5-bis(1, 1-Dimethylethyl)phenyl]-3-oxo-1-propenyl]benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

202010

 
r & d systems
RDS202010
10mg Each for $192.67

202050

 
r & d systems
RDS202050
50mg Each for $812.12
Description & Specifications

Specifications

Formula Weight 364.47
Quantity 10mg
Chemical Name or Material Ch 55
Assay Percent Range >98%
Molecular Formula C24H28O3
CAS 110368-33-7
Synonym 4-[(1E)-3-[3, 5-bis(1, 1-Dimethylethyl)phenyl]-3-oxo-1-propenyl]benzoic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature