Please login to your online account to display your discounted pricing

CH 223191, Tocris™

Potent aryl hydrocarbon receptor (AhR) antagonist

$195.49 - $816.90

Chemical Identifiers

Chemical Name or Material CH 223191
Assay Percent Range >98%
Molecular Formula C19H19N5O
Formula Weight 333.39
Synonym 1-Methyl-N-[2-methyl-4-[2-(2-methylphenyl)diazenyl]phenyl-1H-pyrazole-5-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

385810

 
r & d systems
385810
10mg Each for $195.49

385850

 
r & d systems
385850
50mg Each for $816.90
Description & Specifications

Specifications

Formula Weight 333.39
Quantity 10mg
Chemical Name or Material CH 223191
Synonym 1-Methyl-N-[2-methyl-4-[2-(2-methylphenyl)diazenyl]phenyl-1H-pyrazole-5-carboxamide
CAS 301326-22-7
Assay Percent Range >98%
Molecular Formula C19H19N5O
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C