Please login to your online account to display your discounted pricing

CGS 21680 hydrochloride, Tocris


A2A agonist

Quantity 10mg

Manufacturer: r & d systems  106310

 View more versions of this product

Catalog No. 10-631-0

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material CGS 21680 hydrochloride
CAS 124431-80-7
Recommended Storage Desiccate at -20°C
Quantity 10mg
Assay Percent Range >98%
Molecular Formula C23H29N7O6·HCl
Formula Weight 535.99
Synonym 4-[2-[[6-Amino-9-(N-ethyl-β-D-ribofuranuronamidosyl)-9H-purin-2-yl]amino]ethyl]benzenepropanoic acid hydrochloride

Specifications

Chemical Name or Material CGS 21680 hydrochloride
Assay Percent Range >98%
Molecular Formula C23H29N7O6·HCl
Formula Weight 535.99
Synonym 4-[2-[[6-Amino-9-(N-ethyl-β-D-ribofuranuronamidosyl)-9H-purin-2-yl]amino]ethyl]benzenepropanoic acid hydrochloride
Accessories

Desiccate at -20°C